Produktname
3-Hydroxybuttersäure analytical standard
Produktnummer
52017
Produktmarke
sial
CAS-Nummer
300-85-6
Molekulargewicht
104.10
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless to Light Yellow |
| APPEARANCE (FORM) | Liquid or Solid |
| PURITY (GC AREA %) | ≥ 98.0 % |
| REFRACTIVE INDEX N20/D | 1.439 - 1.445 |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |