Produktname
2-Hydroxy-1-naphthaldehydoxim ≥98.0% (HPLC)
Produktnummer
97024
Produktmarke
sial
CAS-Nummer
7470-09-9
Molekulargewicht
187.19
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Red |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| CARBON CONTENT | 70.58 % (THEORY) |
| HYDROGEN CONTENT | 4.85 % (THEORY) |
| NITROGEN CONTENT | 7.48 % (THEORY) |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |