Produktname
Atto 647N BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Produktnummer
04507
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Report Result |
| APPEARANCE (FORM) | Report Result |
| PURITY (HPLC AREA %) | ≥ 90.0 % |
| Absorption Max | 640.0 - 646.0 nm |
| SOLVENT (UV) | ethanol with 0.1% trifluoroacetic acid |