Produktname
D-Sedoheptulose ≥95% (TLC)
Produktnummer
07532
Produktmarke
sigma
CAS-Nummer
3019-74-7
Molekulargewicht
210.18
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Beige to Very Dark Beige and Brown to Very Dark Brown and Red-Brown and Very Dark Red-Brown |
| APPEARANCE (FORM) | Solid or Semi-Solid or Viscous Liquid |
| PURITY (TLC AREA %) | ≥ 95 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| CARBON 13 NMR SPECTRUM | CONFORMS TO STRUCTURE |