Produktname
Abberior® STAR 635P, NHS-Ester suitable for fluorescence
Produktnummer
07679
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | REPORT RESULT |
| APPEARANCE (FORM) | Powder or Solid or Crystals |
| SOLUBILITY (COLOR) | REPORT RESULT |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 0.25 MG/ML IN DMF |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| DEGREE OF COUPLING | ≥ 80.0 % |
| EMISSION MAXIMUM | 654 ± 10 nm |
| REMARKS ON LUMINESCENCE | EXC.638 nm IN PBS PH7.4 |
| RECOMMENDED RETEST PERIOD | 24 MONTHS |