Produktname
α-Amylase aus Bacillus sp. powder, yellow-brown, ~50 U/mg
Produktnummer
10070
Produktmarke
sigma
CAS-Nummer
9000-90-2
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Brown |
| APPEARANCE (FORM) | Powder |
| SOLUBILITY (COLOR) | Colorless to Faint Brown |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 5 MG/ML 23 MM K PHOSPHATE, 6.6 MM NACL, PH 6.9 |
| ENZYMATIC ACTIVITY | 40.0 - 60.0 U/mg |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |