Produktname
Histopaque®-1077 sterile-filtered, density: 1.077
Produktnummer
10771
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Appearance (Turbidity) | Clear |
| Appearance (Form) | Liquid |
| Appearance (Colour) | Colorless |
| pH | 8.8 - 9.0 |
| Osmolality | - mOs/kg |
| Sterility | Pass |
| Endotoxin Level | <= 3 EU/ml |
| Density at 25°C | 1.076 - 1.078 g/cm3 |
| Performance | Conforms |
| Opaque Layer Present | Pass |
| %Mononuclear cells of total cell count | >= 90 % |
| % Viability | >= 85 % |