Produktname
1-Phenyl-1,3-butandion purum, ≥98.0% (GC)
Produktnummer
12830
Produktmarke
sigma
CAS-Nummer
93-91-4
Molekulargewicht
162.19
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Dark Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 98.0 % |
| MELTING POINT | 54 - 56 C |
| SOLUBILITY (COLOR) | Colorless to Light Yellow |
| SOLUBILITY (TURBIDITY) | Clear to Hazy |
| SOLUBILITY (METHOD) | 1G IN 10ML DIOXANE |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |