Produktname
Glucosedehydrogenase aus Pseudomonas sp. powder, white, ≥200 U/mg
Produktnummer
19359
Produktmarke
sigma
CAS-Nummer
9028-53-9
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | -- |
| ENZYMATIC ACTIVITY | ≥ 200 U/mg |
| UNIT DEFINITION | ONE UNIT CORRESPONDS TO THE AMOUNT OF ENZYME WHICH OXIDIZES 1 UMOLE B-D-GLUCOSE TO D-GLUCONO-D-LACTONE PER MINUTE AT PH 8.0 AND 37 DEG C |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |