Produktname
5(6)-Carboxy-2′,7′-dichlorfluoreszeindiacetat BioReagent, suitable for fluorescence, ≥85% (HPCE)
Produktnummer
21884
Produktmarke
sigma
CAS-Nummer
127770-45-0
Molekulargewicht
529.28
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow and White to Light Orange |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPCE AREA %) | ≥ 85.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EMISSION MAXIMUM | 524 - 534 nm |
| REMARKS ON LUMINESCENCE | EX. 470 NM IN 0.1 M TRIS PH 8.0, ESTERASE |
| SUITABILITY ASSAY | SUITABLE AS A FLUOROGENIC SUBSTRATE FOR ESTERASES |