Produktname
Crotonoyl-Coenzym A Trilithiumsalz ~90% (HPLC)
Produktnummer
28007
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
853.41
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Fibers |
| PURITY (HPLC AREA %) | ≥ 85 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 50MG/ML WATER |
| WATER | ≤ 10 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |