Produktname
Alcalase® CLEA ≥5 U/g
Produktnummer
30079
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Brown |
| APPEARANCE (FORM) | Powder or Crystals |
| ENZYMATIC ACTIVITY | ≥5 U/G |
| UNIT DEFINITION | 1 U corresponds to the amount of enzyme which sets free 1 ?mol folin-positive amino acids and peptides (as tyrosine) per minute at pH 8.0 and 37?C (casein as substrate) |