Produktname
Dextran aus Leuconostoc mesenteroides Mr ~200,000
Produktnummer
31398
Produktmarke
sigma
CAS-Nummer
9004-54-0
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White |
| APPEARANCE (FORM) | Powder |
| Molecular Weight Mw (GPC-Analysis) | 170000 - 230000 (GPC) |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear to Hazy |
| SOLUBILITY (METHOD) | 1.5G IN 10ML WATER (30 MIN) |
| LOSS ON DRYING | ≤ 7.0 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |