Produktname
Lugol according to Lugol
Produktnummer
32922
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | REPORT RESULT |
| APPEARANCE (FORM) | REPORT RESULT |
| ARGENTOMETR. TITRATION | ASSAY POTASSIUM IODIDE (KI) |
| REDOX TITRATION | ASSAY IODINE (I) |
| REDOXTITRATION (METHOD) | TITRATION WITH SODIUM THIOSULFATE |
| DENSITY D20/4 | REPORT RESULT |
| RECOMMENDED RETEST PERIOD | 42 MONTHS |