Produktname
Terbuthylazin PESTANAL®, analytical standard
Produktnummer
45678
Produktmarke
sigma
CAS-Nummer
5915-41-3
Molekulargewicht
229.71
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 98.0 % |
| MELTING POINT | 174 - 179 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |