Produktname
Fluroxypyr PESTANAL®, analytical standard
Produktnummer
45758
Produktmarke
sigma
CAS-Nummer
69377-81-7
Molekulargewicht
255.03
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow to Faint Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| MELTING POINT | 214 - 228 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |