Produktname
Esterase aus Pferdeleber lyophilized, powder, brown, 0.5-1.0 U/mg
Produktnummer
46069
Produktmarke
sigma
CAS-Nummer
9016-18-6
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Faint Brown to Dark Brown |
| APPEARANCE (FORM) | Powder |
| SOLUBILITY (COLOR) | Faint Brown to Brown |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 5 MG/ML H2O |
| ENZYMATIC ACTIVITY | 0.50 - 1.00 U/mg |
| RECOMMENDED RETEST PERIOD | 42 MONTHS |