Produktname
Ofurace PESTANAL®, analytical standard
Produktnummer
46143
Produktmarke
sigma
CAS-Nummer
58810-48-3
Molekulargewicht
281.73
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow to Faint Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 98.0 % |
| MELTING POINT | 143 - 148 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |