Produktname
Formiatdehydrogenase aus Candida boidinii liquid (clear), clear brown, 40.0-60.0 U/mL
Produktnummer
47753
Produktmarke
sigma
CAS-Nummer
9028-85-7
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Very Light Brown to Dark Brown |
| APPEARANCE (FORM) | Liquid |
| APPEARANCE (TURBIDITY) | Slightly Hazy to Turbid |
| SOLUBILITY (COLOR) | Colorless to Light Brown |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 1:10 IN H2O |
| ENZYMATIC ACTIVITY | 40.0 - 60.0 U/ml |
| UNIT DEFINITION | 1 U CORRESPONDS TO THE AMOUNT OF ENZYME WHICH OXIDIZES 1 UMOL SODIUM FORMATE (CAT. NO. 71539) PER MINUTE AT PH 7.6 AND 25 DEG C. |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |