Produktname
Phalloidin-Atto 488 BioReagent, suitable for fluorescence, ≥90% (HPLC)
Produktnummer
49409
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Report Result |
| APPEARANCE (FORM) | Report Result |
| PURITY (HPLC AREA %) | ≥ 90 % |
| Absorption Max | 499 - 505 nm |
| SOLVENT (UV) | water |