Produktname
PureCol™ Collagen powder from Bovine hide, 97% Type I with the remainder of Type III collagen, suitable for cell culture
Produktnummer
5006
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Sterility (USP modified) | No Growth |
| Endotoxin LAL (EU/mL) | ≤ 1.0 |
| Purity - SDS Page Silver Stain | ≥ 99.9% |
| Electrophoretic Pattern | Characteristic |