Produktname
VitroCol® Collagen from Human neonatal fibroblasts, 97% Type I with the remainder of Type III collagen, suitable for cell culture
Produktnummer
5008
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Sterility | No growth |
| Endotoxin Level | <1.0 EU/mL |
| Electrophoresis | > 85% collagen containing within alpha, beta, and gamma band, < 15% collagen contained within bands traveling faster than alpha |