Produktname
Kollagen from bovine flexor tendon
Produktnummer
5162
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Appearance | Report result |
| Purity | > 96% |
| Endotoxin | < 20 EU/gram |
| Bioburden (CFU/mL) | < 200 cfu/gram |
Test | Spezifikation |
|---|---|
| Appearance | Report result |
| Purity | > 96% |
| Endotoxin | < 20 EU/gram |
| Bioburden (CFU/mL) | < 200 cfu/gram |