Produktname
Robinin ≥90.0% (HPLC)
Produktnummer
53609
Produktmarke
sigma
CAS-Nummer
301-19-9
Molekulargewicht
740.66
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow |
| APPEARANCE (FORM) | Powder |
| PURITY (HPLC AREA %) | ≥ 90.0 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |