Produktname
Kaliumbicarbonat BioUltra, ≥99.5% (T)
Produktnummer
60339
Produktmarke
sigma
CAS-Nummer
298-14-6
Molekulargewicht
100.12
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals |
| TITRATION (T) HCL 1M | 99.5 - 101.0 % |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| ALUMINIUM (ICP) | ≤ 5 mg/kg |
| BARIUM (ICP) | ≤ 5 mg/kg |
| BISMUTH (ICP) | ≤ 5 mg/kg |
| CALCIUM (ICP) | ≤ 10 mg/kg |
| CADMIUM (ICP) | ≤ 5 mg/kg |
| COBALT (ICP) | ≤ 5 mg/kg |
| CHROMIUM (ICP) | ≤ 5 mg/kg |
| COPPER (ICP) | ≤ 5 mg/kg |
| IRON (ICP) | ≤ 5 mg/kg |
| LITHIUM (ICP) | ≤ 5 mg/kg |
| MAGNESIUM (ICP) | ≤ 5 mg/kg |
| MANGANESE (ICP) | ≤ 5 mg/kg |
| MOLYBDENUM (ICP) | ≤ 5 mg/kg |
| SODIUM (ICP) | ≤ 200 mg/kg |
| NICKEL (ICP) | ≤ 5 mg/kg |
| LEAD (ICP) | ≤ 5 mg/kg |
| STRONTIUM (ICP) | ≤ 5 mg/kg |
| ZINC (ICP) | ≤ 5 mg/kg |
| ARSENIC TRACES (MHS-AAS) | ≤ 1 mg/kg |
| TOTAL SULFUR AS SO4 (ICP) | ≤ 50 mg/kg |
| CHLORIDE (CL) | ≤ 50 mg/kg |
| SOLUBILITY (METHOD) | 1M in water |
| APPEARANCE (SOLUTION) | CLEAR COLORLESS |
| PH (SOLUTION) | 7.5 - 9.0 |
| RESIDUE (FILTER TEST) | NO RESIDUE |
| UV - ABS. AT 260 NM | ≤ 0.01 |
| UV - ABS. AT 280 NM | ≤ 0.01 |
| RECOMMENDED RETEST PERIOD | 72 MONTHS |