Produktname
Gitoxin ≥99% (HPLC)
Produktnummer
64107
Produktmarke
sigma
CAS-Nummer
4562-36-1
Molekulargewicht
780.94
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 99 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| RECOMMENDED RETEST PERIOD | 72 MONTHS |