Produktname
Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranosid ≥97.0% (TLC)
Produktnummer
67328
Produktmarke
sigma
CAS-Nummer
115457-83-5
Molekulargewicht
335.39
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (TLC AREA %) | ≥ 99.0 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 50MG IN 5ML WATER |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| CALCIUM (ICP) | ≤ 50 mg/kg |
| CADMIUM (ICP) | ≤ 50 mg/kg |
| COBALT (ICP) | ≤ 50 mg/kg |
| CHROMIUM (ICP) | ≤ 50 mg/kg |
| COPPER (ICP) | ≤ 50 mg/kg |
| IRON (ICP) | ≤ 50 mg/kg |
| POTASSIUM (ICP) | ≤ 50 mg/kg |
| MAGNESIUM (ICP) | ≤ 50 mg/kg |
| MANGANESE (ICP) | ≤ 50 mg/kg |
| SODIUM (ICP) | ≤ 1000 mg/kg |
| NICKEL (ICP) | ≤ 50 mg/kg |
| LEAD (ICP) | ≤ 50 mg/kg |
| ZINC (ICP) | ≤ 50 mg/kg |