Produktname
Natriumpersulfat BioUltra, ≥99%
Produktnummer
71889
Produktmarke
sigma
CAS-Nummer
7775-27-1
Molekulargewicht
238.10
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals |
| REDOX TITRATION | 99 - 101 % |
| REDOXTITRATION (METHOD) | IODOMETRIC |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| ALUMINIUM (ICP) | ≤ 5 mg/kg |
| BARIUM (ICP) | ≤ 5 mg/kg |
| BISMUTH (ICP) | ≤ 5 mg/kg |
| CALCIUM (ICP) | ≤ 50 mg/kg |
| CADMIUM (ICP) | ≤ 5 mg/kg |
| COBALT (ICP) | ≤ 5 mg/kg |
| CHROMIUM (ICP) | ≤ 5 mg/kg |
| COPPER (ICP) | ≤ 5 mg/kg |
| IRON (ICP) | ≤ 5 mg/kg |
| LITHIUM (ICP) | ≤ 5 mg/kg |
| MAGNESIUM (ICP) | ≤ 5 mg/kg |
| MANGANESE (ICP) | ≤ 5 mg/kg |
| MOLYBDENUM (ICP) | ≤ 5 mg/kg |
| NICKEL (ICP) | ≤ 5 mg/kg |
| LEAD (ICP) | ≤ 5 mg/kg |
| STRONTIUM (ICP) | ≤ 5 mg/kg |
| ZINC (ICP) | ≤ 5 mg/kg |
| ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
| CHLORIDE (CL) | ≤ 50 mg/kg |
| SOLUBILITY (METHOD) | 1M IN H2O |
| APPEARANCE (SOLUTION) | CLEAR COLORLESS |
| PH (SOLUTION) | 2.5 - 4.0 |
| RESIDUE (FILTER TEST) | NO RESIDUE |
| CUT-OFF | ≤ 309 nm |
| RECOMMENDED RETEST PERIOD | 72 MONTHS |