Produktname
L-Ribulose ≥90% (HPLC)
Produktnummer
75436
Produktmarke
sigma
CAS-Nummer
2042-27-5
Molekulargewicht
150.13
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless to Yellow |
| APPEARANCE (FORM) | Liquid or Viscous Liquid |
| PURITY (HPLC AREA %) | ≥ 90 % |
| SPECIFIC ROTATION (20/D) | 14.5 ± 3.0 Degrees |
| CONCENTRATION | C=0.1 IN WATER |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |