Produktname
Hyaluronsäure Natriumsalz aus Streptococcus equi sp. mol wt 500,000-750,000
Produktnummer
75574
Produktmarke
sigma
CAS-Nummer
9067-32-7
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White |
| APPEARANCE (FORM) | Powder |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| AAS QUALITATIVE | CONFIRM SODIUM |
| SUITABILITY ASSAY | suitable as substrate for hyaluronidase |