Produktname
Anti-Maus-IgG – Atto 594 in Ziege hergestellte Antikörper ~1 mg/mL protein, affinity isolated antibody
Produktnummer
76085
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Light Purple to Dark Purple to Dark Blue |
| APPEARANCE (FORM) | Liquid |
| EMISSION MAXIMUM | 625 ± 5 nm |
| REMARKS ON LUMINESCENCE | EXC. 607 NM IN PBS |
| PROTEIN CONTENT (280 NM) | ≥ 0.8 mg/ml |
| DYE-TO-PROTEIN RATIO (MOLAR) | ≥ 2.0 |
| SDS GEL ELECTROPHORESIS | CORRESPONDS TO REQUIREMENTS |
| REM. ON GELELECTROPHORESIS | UNCONJUGATED DYE ≤ 5 % OF TOTAL FLUORESCENCE |
| RECOMMENDED RETEST PERIOD | 24 MONTHS |