Produktname
Pentacen suitable for fluorescence, >95.0%
Produktnummer
76440
Produktmarke
sigma
CAS-Nummer
135-48-8
Molekulargewicht
278.35
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Dark Blue to Very Dark Blue and Dark Purple to Very Dark Purple and Black |
| APPEARANCE (FORM) | Powder or Crystals |
| CARBON CONTENT | 90.2 - 99.7 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |