Produktname
Anti-Kaninchen-IgG – Atto 594 in Ziege hergestellte Antikörper
Produktnummer
77671
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Blue to Dark Blue and Purple to Dark Purple and Red to Dark Red |
| APPEARANCE (FORM) | Liquid |
| EMISSION MAXIMUM | 626.0 ± 5.0 nm |
| REMARKS ON LUMINESCENCE | EXC. 607 NM IN PBS |
| PROTEIN CONTENT (280 NM) | ≥ 0.8 mg/ml |
| DYE-TO-PROTEIN RATIO (MOLAR) | ≥ 2.0 |
| SDS GEL ELECTROPHORESIS | CORRESPONDS TO REQUIREMENTS |
| REM. ON GELELECTROPHORESIS | UNCONJUGATED DYE ≤ 5 % OF TOTAL FLUORESCENCE |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |