Produktname
Phalloidin-Atto 700 suitable for fluorescence, ≥90% (HPLC)
Produktnummer
79286
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Report Result |
| APPEARANCE (FORM) | Report Result |
| PURITY (HPLC AREA %) | ≥ 90.0 % |
| Absorption Max | 687 - 693 nm |
| SOLVENT (UV) | methanol |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |