Produktname
1,6-Hexandiol -Lösung BioUltra, ~6 M in H2O
Produktnummer
88571
Produktmarke
sigma
CAS-Nummer
629-11-8
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| DENSITY D20/4 | 0.975 - 1.104 |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| ALUMINIUM (ICP) | ≤ 5.0 mg/kg |
| BARIUM (ICP) | ≤ 5.0 mg/kg |
| BISMUTH (ICP) | ≤ 5.0 mg/kg |
| CALCIUM (ICP) | ≤ 10 mg/kg |
| CADMIUM (ICP) | ≤ 5.0 mg/kg |
| COBALT (ICP) | ≤ 5.0 mg/kg |
| CHROMIUM (ICP) | ≤ 5.0 mg/kg |
| COPPER (ICP) | ≤ 5.0 mg/kg |
| IRON (ICP) | ≤ 5.0 mg/kg |
| POTASSIUM (ICP) | ≤ 200 mg/kg |
| LITHIUM (ICP) | ≤ 5.0 mg/kg |
| MAGNESIUM (ICP) | ≤ 5.0 mg/kg |
| MANGANESE (ICP) | ≤ 5.0 mg/kg |
| MOLYBDENUM (ICP) | ≤ 5.0 mg/kg |
| SODIUM (ICP) | ≤ 200 mg/kg |
| NICKEL (ICP) | ≤ 5.0 mg/kg |
| LEAD (ICP) | ≤ 5.0 mg/kg |
| STRONTIUM (ICP) | ≤ 5.0 mg/kg |
| ZINC (ICP) | ≤ 5.0 mg/kg |
| RESIDUE (FILTER TEST) | NO RESIDUE |
| UV - ABS. AT 260 NM | ≤ 0.1 |
| UV - ABS. AT 280 NM | ≤ 0.1 |