Produktname
Cacodylat pH 7.0; 0.05 M – Magnesiumchlorid 9 mM – Spermin 2.25 mM – Kobalt(III)-hexamin 1.8 mM – Spermidin 0.9 mM – PEG 400 5%
Produktnummer
89269
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless to Very Light Brown |
| APPEARANCE (FORM) | Liquid |
| DENSITY D20/4 | 1.011 - 1.015 |
| REFRACTIVE INDEX N20/D | 1.341 - 1.343 |
| PH | 6.8 - 7.2 |