Produktname
Polyacrylamid nonionic water-soluble polymer
Produktnummer
92560
Produktmarke
sigma
CAS-Nummer
9003-05-8
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| VISCOSITY (ROTATION) | 6 - 15 mPas |
| VISCOSITY (CONDITIONS) | 0.5% in H2O at 20 C, DIN 45, 100 s-1 |