Produktname
Hoechst 33258 -Lösung 1 mg/mL in H2O, ≥98.0% (HPLC)
Produktnummer
94403
Produktmarke
sigma
CAS-Nummer
23491-45-4
Molekulargewicht
533.88
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Very Light Yellow to Dark Yellow and Very Light Green-Yellow to Dark Green-Yellow |
| APPEARANCE (FORM) | Liquid |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| REFRACTIVE INDEX N20/D | 1.332 - 1.334 |
| LUMINESCENCE | CORRESPONDS TO REQUIREMENTS |
| EMISSION MAXIMUM | 460.0 - 470.0 nm |