Produktname
p-Xylenolphthalein
Produktnummer
95620
Produktmarke
sigma
CAS-Nummer
50984-88-8
Molekulargewicht
374.43
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Beige to Faint Orange |
| APPEARANCE (FORM) | Powder |
| THIN LAYER CHROMATOGR. | CORRESPONDS TO STANDARD |
| SOLUBILITY (COLOR) | Colorless to Light Yellow and Colorless to Light Green-Yellow |
| SOLUBILITY (METHOD) | 0.01G IN 10ML ETHANOL |
| CARBON CONTENT | 76.99 % (THEORY) |
| HYDROGEN CONTENT | 5.92 % (THEORY) |
| WAVELENGTH (1) (UV) | 587 - 597 nm |
| EXTINCTION (1%/1CM) (1) (UV) | ≥ 800 |
| SOLVENT (UV) | WATER (ALKALINE) |