Produktname
(+)-6-Aminopenicillansäure
Produktnummer
a1005
Produktmarke
sigma
CAS-Nummer
551-16-6
Molekulargewicht
216.26
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO WHITE WITH A DARK YELLOW CAST POWDER |
| SOLUBILITY | CLEAR TO VERY SLIGHTLY HAZY COLORLESS TO LIGHT YELLOW SOLUTION AT 50MG/ML IN 1M HYDROCHLORIC ACID |
| IR SPECTRUM | CONSISTENT WITH STRUCTURE |
| ELEMENTAL ANALYSIS | 43.5 TO 45.3%CARBON |
| 12.7 TO 13.3%NITROGEN | |
| APPROVED JLH 10/20/95 |