Produktname
4-Amino-hippursäure ≥99%
Produktnummer
a1422
Produktmarke
sigma
CAS-Nummer
61-78-9
Molekulargewicht
194.19
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow and Faint Beige to Light Beige and Faint Brown to Light Brown |
| APPEARANCE (FORM) | Powder |
| TITRATION (T) NAOH 0.1M | ≥99 % |
| SOLUBILITY (COLOR) | Colorless to Yellow |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 50MG/ML IN WATER |