Produktname
Ala-Ala-Ala-Ala-Ala
Produktnummer
a5025
Produktmarke
sigma
CAS-Nummer
10183-34-3
Molekulargewicht
373.40
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO OFF-WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS TO LIGHT YELLOW SOLUTION AT 50 MG/ML IN 1 M 1 M HYDROCHLORIC ACID |
| IDENTITY | CONSISTENT WITH STRUCTURE BY IR OR CONSISTENT WITH STRUCT BY NMR,H |
| WATER CONTENT BY KARL FISCHER | < OR = 6.0% |
| PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 99% |
| DOCUMENT # | A5025/05/12/08/1 |