Produktname
Acetylthiocholinchlorid ≥99% (TLC), powder
Produktnummer
a5626
Produktmarke
sigma
CAS-Nummer
6050-81-3
Molekulargewicht
197.73
Test | Spezifikation |
|---|---|
| Appearance (Color) | White to Off-White |
| Appearance (Form) | Powder |
| Solubility (Color) | Colorless |
| Solubility (Turbidity) | Clear to Slightly Hazy |
| 100 mg/mL in H2O | |
| Infrared spectrum | Conforms to Structure |
| Purity (TLC) | ≥99 % |
| Suitability | Suitable |
| Enzymatic Suitability; Suitable as substrate for | |
| Cholinesterase. |