Produktname
Ammoniummetavanadat suitable for cell culture, powder
Produktnummer
a8175
Produktmarke
sigma
CAS-Nummer
7803-55-6
Molekulargewicht
116.98
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO LIGHT YELLOW POWDER |
| SOLUBILITY | CLEAR YELLOW TO YELLOW-GREEN SOLUTION AT 25 MG PLUS 5 ML WATER |
| PURITY BY REDOX TITRATION | NLT 99% |
| CELL CULTURE TEST | PASSES |
| DOCUMENT # A8175/2/96/1 |