Produktname
Antide ≥97% (HPLC)
Produktnummer
a8802
Produktmarke
sigma
CAS-Nummer
112568-12-4
Molekulargewicht
1591.29
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS SOLUTION AT 1 MG/ML IN WATER |
| PEPTIDE CONTENT BY AMINO ACID ANALYSIS | > OR = 80% |
| PURITY BY HPLC | > OR = 97% |
| DOCUMENT # | A8802/12/11/09/1 |