Produktname
Antithrombin III aus Rinderplasma lyophilized powder, 200-400 units/mg protein (E1%/280 = 6.5)
Produktnummer
a9141
Produktmarke
sigma
CAS-Nummer
90170-80-2
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS SOLUTION AT 10 UNITS PLUS 1 ML OF WATER |
| SPECIFIC ACTIVITY | 200 TO 400 UNITS/MG PROTEIN |
| UNIT DEFINITION | ONE UNIT IS THE ACTIVITY PRESENT IN 0.1 ML OF NORMAL HUMAN POOLED PLASMA TESTED IN THE PRESENCE OF 0.1 UNIT OF HEPARIN. |
| APPROVED HAM 10/21/91 |