Produktname
DL-Arabinose ≥98% (GC)
Produktnummer
a9524
Produktmarke
sigma
CAS-Nummer
147-81-9
Molekulargewicht
150.13
Test | Spezifikation |
|---|---|
| Appearance (Color) | White to Off-White |
| Appearance (Form) | Powder |
| Solubility (Color) | Colorless |
| Solubility (Turbidity) | Clear |
| 50 mg/ml, H2O | |
| Purity (GC) | ≥98 % |