Produktname
Anti-KHK antibody produced in rabbit affinity isolated antibody
Produktnummer
av41376
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Concentration | 0.5-1 mg/ml |
| ELISA | Not Verified |
| Western Blotting | Antibody dilution: 1 µg/ml |
| Immunohistochemistry | Not Verified |
| Immunofluorescence | Not Verified |
| Immunoprecipitation | Not Verified |
| Flow Cytometry | Not Verified |
| Chromatin Immunoprecipitation | Not Verified |