Produktname
Bradykinin -acetat (Salz) powder, ≥98% (HPLC)
Produktnummer
b3259
Produktmarke
sigma
CAS-Nummer
6846-03-3
Molekulargewicht
1060.21 (free base basis)
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White |
| APPEARANCE (FORM) | Powder |
| PURITY (HPLC AREA %) | ≥ 98 % |
| PEPTIDE CONTENT (AMINO ACID ANALYSIS) | ≥ 75 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 25 mg/ml, 0.1 M HOAc |
| WATER | ≤ 10 % |