Produktname
5-Brom-4-Chlor-3-Indolylphosphat Dinatriumsalz ≥98% (HPLC)
Produktnummer
b6149
Produktmarke
sigma
CAS-Nummer
102185-33-1
Molekulargewicht
370.43
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Blue |
| APPEARANCE (FORM) | Powder |
| PURITY (HPLC AREA %) | ≥ 98 % |
| SOLUBILITY (COLOR) | COLORLESS TO LIGHT YELLOW TO LIGHT BLUE |
| SOLUBILITY (TURBIDITY) | Clear to Hazy |
| SOLUBILITY (METHOD) | 50MG/ML IN WATER |
| WATER | ≤ 10 % |
| CARBON CONTENT | 25.0 - 26.4 % (ANHYDROUS BASIS) |
| NITROGEN CONTENT | 3.5 - 4.1 % (ANHYDROUS BASIS) |
| SODIUM | 11.5 - 14.0 % (ANHYDROUS BASIS) |
| Phosphorus | 7.8 - 9.0 % (ANHYDROUS BASIS) |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |